Learn More
TRAIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10332-1-AP-20UL
 View more versions of this product
Product Code. 16866653
  • € 145.00 / 20µL
Description
TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.
Specifications
Specifications
TRAIP
Polyclonal
Unconjugated
TRAIP
RING finger protein 206, RNF206, TRAF interacting protein, TRAIP, TRIP
Rabbit
Antigen Affinity Chromatography
RUO
10293, 22036
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8VIG6, Q9BWF2
TRAIP
TRAIP Fusion Protein Ag0491
20 μL
Primary
Human, Mouse