missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10332-1-AP-150UL
This item is not returnable.
View return policy
Description
TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.Specifications
| TRAIP | |
| Polyclonal | |
| Unconjugated | |
| TRAIP | |
| RING finger protein 206, RNF206, TRAF interacting protein, TRAIP, TRIP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10293, 22036 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8VIG6, Q9BWF2 | |
| TRAIP | |
| TRAIP Fusion Protein Ag0491 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction