missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRAM2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13250879
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13250879 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13250879 Supplier Invitrogen™ Supplier No. PA525887

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRAM2 Polyclonal Antibody for Western Blot, IHC (P)

Necessary for collagen type I synthesis. May couple the activity of the ER Ca(2+) pump SERCA2B with the activity of the translocon. This coupling may increase the local Ca(2+) concentration at the site of collagen synthesis, and a high Ca(2+) concentration may be necessary for the function of molecular chaperones involved in collagen folding. Required for proper insertion of the first transmembrane helix N-terminus of TM4SF20 into the ER lumen, may act as a ceramide sensor for regulated alternative translocation (RAT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TRAM2
Gene Accession No. Q15035
Gene Alias C330003D03Rik; KIAA0057; LOC100361830; tram2; TRAM-like protein; Translocating chain-associated membrane protein 2; translocating chain-associating membrane protein 2; translocation associated membrane protein 2; wu:fb20c11; wu:fi23a12; zgc:111902; zgc:55869; zgc:76937
Gene Symbols TRAM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 342-370 amino acids from the C-terminal region of human TRAM2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9697
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.