missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody (12A668), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56512DL594
This item is not returnable.
View return policy
Description
TRANCE/TNFSF11/RANK L Monoclonal antibody specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, Flow (Cell Surface), ChIP assay (ChIP)
Specifications
| TRANCE/TNFSF11/RANK L | |
| Monoclonal | |
| DyLight 594 | |
| CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE/TNFSF11/RANK L was used as immunogen. | |
| 0.1 mL | |
| Cancer | |
| 8600 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 12A668 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction