missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody (685857), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6264PECY7
This item is not returnable.
View return policy
Description
TRANCE/TNFSF11/RANK L Monoclonal antibody specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for Flow Cytometry
Specifications
| TRANCE/TNFSF11/RANK L | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 685857 | |
| Flow Cytometry | |
| CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| Mouse myeloma cell line NS0-derived recombinant human TRANCE/TNFSF11/RANK L, Gly136-Asp317, Accession # O14788 | |
| 0.1 mL | |
| Cancer | |
| 8600 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction