missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trandolapril, MedChemExpress
Description
Trandolapril (RU44570) is a nonsulfhydryl prodrug that is hydrolysed to the active diacid Trandolaprilat. Trandolapril is an orally administered angiotensin converting enzyme (ACE) inhibitor that has been used in the treatment of hypertension and congestive heart failure (CHF), and after myocardial infarction (MI).
Specifications
Specifications
| Chemical Name or Material | Trandolapril |
| CAS | 87679-37-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H34N2O5 |
| Quantity | 50 mg |
| Synonym | RU44570 |
| Solubility Information | DMSO : 100 mg/mL (232.27 mM; Need ultrasonic) |
| SMILES | O=C([C@H]1N(C([C@@H](N[C@H](C(OCC)=O)CCC2=CC=CC=C2)C)=O)[C@@]3([H])CCCC[C@]3([H])C1)O |
| Molecular Weight (g/mol) | 430.54 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?