missing translation for 'onlineSavingsMsg'
Learn More

Tranexamic Acid Dimer, TRC

Product Code. 30003308
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003308 25 mg 25mg
30007325 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003308 Supplier Toronto Research Chemicals Supplier No. TRCT71451525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93940-19-3
Molecular Formula C16 H27 N O4
Molecular Weight (g/mol) 297.39
Synonym Cyclohexanecarboxylic acid, 4,4'-[iminobis(methylene)]bis-, [trans(trans)]- (9CI), trans,trans-4,4'-(Iminodimethylene)di(cyclohexanecarboxylic acid), Tranexamic acid dimer, trans-trans-4,4'-Iminodimethylenedi(cyclohexanecarboxylic acid), (trans,trans)-4,4'-(Azanediylbis(methylene))dicyclohexanecarboxylic acid, 4-[[(4-Carboxycyclohexyl)methylamino]methyl]cyclohexane-1-carboxylic acid, Tranexamic acid EP Imp. A
IUPAC Name 4-[[(4-carboxycyclohexyl)methylamino]methyl]cyclohexane-1-carboxylic acid
SMILES OC(=O)[C@@H]1CC[C@@H](CNC[C@@H]2CC[C@H](CC2)C(=O)O)CC1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H27NO4/c18-15(19)13-5-1-11(2-6-13)9-17-10-12-3-7-14(8-4-12)16(20)21/h11-14,17H,1-10H2,(H,18,19)(H,20,21)/t11-,12-,13-,14-
Formula Weight 297.194
Percent Purity >95%
Chemical Name or Material Tranexamic acid dimer
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.