missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2,5-Dimethylpiperazine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | trans-2,5-Dimethylpiperazine |
| CAS | 2815-34-1 |
| Molecular Formula | C6H14N2 |
| Quantity | 1 g |
| Synonym | (2R,5S)-2,5-Dimethylpiperazine, NSC 3708, meso-2,5-Dimethylpiperazine |
| InChI Formula | InChI=1S/C6H14N2/c1-5-3-8-6(2)4-7-5/h5-8H,3-4H2,1-2H3/t5-,6+ |
| SMILES | C[C@@H]1CN[C@@H](C)CN1 |
| IUPAC Name | (2R,5S)-2,5-dimethylpiperazine |
| Molecular Weight (g/mol) | 114.19 |
| Formula Weight | 114.1157 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction