Learn More
trans-2-Amino-N-(4-hydroxycyclohexyl)benzamide, Mikromol™
Product Code. 30050407
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30050407
Brand: Mikromol MM0005.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
trans-2-Amino-N-(4-hydroxycyclohexyl)benzamide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material trans-2-Amino-N-(4-hydroxycyclohexyl)benzamide
CAS 15942-11-7
Analyte or Component Names trans-2-Amino-N-(4-hydroxycyclohexyl)benzamide
Shipping Condition Room Temperature
Molecular Formula C13 H18 N2 O2
Certifications/Compliance ISO 17025
Synonym trans-2-Amino-N-(4-hydroxycyclohexyl)benzamide, Benzamide, o-amino-N-(4-hydroxycyclohexyl)-, trans- (8CI)
InChI Formula InChI=1S/C13H18N2O2/c14-12-4-2-1-3-11(12)13(17)15-9-5-7-10(16)8-6-9/h1-4,9-10,16H,5-8,14H2,(H,15,17)/t9-,10-
SMILES Nc1ccccc1C(=O)N[C@@H]2CC[C@@H](O)CC2
IUPAC Name 2-amino-N-(4-hydroxycyclohexyl)benzamide
Molecular Weight (g/mol) 234.29
Quantity 100 mg
Formula Weight 234.1368 g/mol