missing translation for 'onlineSavingsMsg'
Learn More

trans-3,4,5,4'-Tetramethoxystilbene, TRC

Product Code. 30413132
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413132 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413132 Supplier Toronto Research Chemicals Supplier No. TRCT298908250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-3,4,5,4'-Tetramethoxystilbene
CAS 134029-62-2
Molecular Formula C18 H20 O4
Quantity 250 mg
Synonym trans-3,4,5,4'-Tetramethoxystilbene
InChI Formula InChI=1S/C18H20O4/c1-19-15-9-7-13(8-10-15)5-6-14-11-16(20-2)18(22-4)17(12-14)21-3/h5-12H,1-4H3/b6-5+
SMILES COc1ccc(\C=C\c2cc(OC)c(OC)c(OC)c2)cc1
IUPAC Name 1,2,3-trimethoxy-5-[(E)-2-(4-methoxyphenyl)ethenyl]benzene
Molecular Weight (g/mol) 300.349
Formula Weight 300.136
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.