Learn More
trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride, Mikromol™
Product Code. 17991446
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991446 25 mg 25mg
17433752 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17991446 Supplier Mikromol Supplier No. MM0005.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride
CAS 15942-08-2
Analyte or Component Names trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H18 Br2 N2 O . Cl H
Certifications/Compliance ISO 17025
Synonym Ambroxol Hydrochloride Imp. B (EP), Ambroxol Imp. B (EP), trans-4-(6,8-Dibromo-1,4-dihydroquinazolin-3(2H)-yl)cyclohexanol Hydrochloride, Ambroxol Hydrochloride Imp. B (EP) as Hydrochloride, trans-4-(6,8-Dibromo-1,4-dihydro-3(2H)-quinazolinyl)cyclohexanol monohydrochloride, Ambroxol Hydrochloride Impurity B as Hydrochloride, Ambroxol Impurity B as Hydrochloride, trans-4-(6,8-Dibromo-1,4-dihydro-3(2H)-quinazolinyl)cyclohexanol Hydrochloride, NA 873 Hydrochloride, Ambroxol EP Impurity B Hydrochloride, Ambroxol Cyclic Impurity Hydrochloride
InChI Formula InChI=1S/C14H18Br2N2O.ClH/c15-10-5-9-7-18(8-17-14(9)13(16)6-10)11-1-3-12(19)4-2-11;/h5-6,11-12,17,19H,1-4,7-8H2;1H/t11-,12-;
SMILES Cl.O[C@@H]1CC[C@H](CC1)N2CNc3c(Br)cc(Br)cc3C2
IUPAC Name 4-(6,8-dibromo-2,4-dihydro-1H-quinazolin-3-yl)cyclohexan-1-ol;hydrochloride
Molecular Weight (g/mol) 426.57
Quantity 25 mg
Formula Weight 423.9553 g/mol