Learn More
trans-ACPD, MedChemExpress
Product Code. 16477616
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477616 100 mg 100mg
16467768 10 mg 10mg
16450316 2 mg 2mg
16433737 50 mg 50mg
16423646 25 mg 25mg
16410238 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16477616 Supplier Medchem Express Supplier No. HY19434
MedChemExpress trans-ACPD, a metabotropic receptor agonist, produces calcium mobilization and an inward current in cultured cerebellar Purkinje neurons.
trans-ACPD, a metabotropic receptor agonist, produces calcium mobilization and an inward current in cultured cerebellar Purkinje neurons.
Specifications
Chemical Name or Material trans-ACPD
Physical Form Solid
CAS 67684-64-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H11NO4
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Trans-(±)-ACP
Solubility Information DMSO : 50 mg/mL (288.73 mM; Need ultrasonic) ∣H2O : 3.57 mg/mL (20.62 mM; Need ultrasonic)
SMILES O=C([C@]1(N)C[C@@H](C(O)=O)CC1)O
Molecular Weight (g/mol) 173.17
Formula Weight 173.17
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only