missing translation for 'onlineSavingsMsg'
Learn More

trans-ACPD, MedChemExpress

Product Code. 16410238
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16410238 5 mg 5mg
16477616 100 mg 100mg
16467768 10 mg 10mg
16450316 2 mg 2mg
16433737 50 mg 50mg
16423646 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16410238 Supplier Medchem Express Supplier No. HY19434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress trans-ACPD, a metabotropic receptor agonist, produces calcium mobilization and an inward current in cultured cerebellar Purkinje neurons.

trans-ACPD, a metabotropic receptor agonist, produces calcium mobilization and an inward current in cultured cerebellar Purkinje neurons.

Specifications

Chemical Name or Material trans-ACPD
Physical Form Solid
CAS 67684-64-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C7H11NO4
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Trans-(±)-ACP
Solubility Information DMSO : 50 mg/mL (288.73 mM; Need ultrasonic) ∣H2O : 3.57 mg/mL (20.62 mM; Need ultrasonic)
SMILES O=C([C@]1(N)C[C@@H](C(O)=O)CC1)O
Molecular Weight (g/mol) 173.17
Formula Weight 173.17
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.