missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transaldolase 1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32216V
This item is not returnable.
View return policy
Description
Transaldolase 1 Polyclonal antibody specifically detects Transaldolase 1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Transaldolase 1 | |
| Polyclonal | |
| Western Blot | |
| dihydroxyacetone transferase, EC 2.2.1.2, glycerone transferase, TAL, TALDO, TALDOR, TALH, TAL-H, transaldolase, transaldolase 1 | |
| The immunogen this antibody was made to, maps to a region between residue 200 to 250 of human Transaldolase 1 using the numbering given in entry NP_006746.1 (GeneID 6888). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 6888 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction