missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transducin alpha Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3504JF646
This item is not returnable.
View return policy
Description
Transducin alpha Polyclonal antibody specifically detects Transducin alpha in Mouse, Porcine, Bovine, Sheep samples. It is validated for Western Blot
Specifications
| Transducin alpha | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse, Pig, Bovine, Sheep | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| CSNBAD3, GBT1, GNATR, guanine nucleotide binding protein (G protein), alpha transducing activitypolypeptide 1, guanine nucleotide-binding protein G(t) subunit alpha-1, guanine nucleotide-binding protein G(T), alpha-1 subunit, rod-type transducin alpha subunit, Transducin alpha-1 chain, transducin, rod-specific | |
| Synthetic Peptide: K(18) L K E D A E K D A R T V K(31) C | |
| 0.1 mL | |
| Signal Transduction, Vision | |
| 2779 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction