missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin Antibody (OTI5G2), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74583APCCY7
This item is not returnable.
View return policy
Description
Transferrin Monoclonal specifically detects Transferrin in Human samples. It is validated for Flow Cytometry.
Specifications
| Transferrin | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| TF | |
| Full length recombinant protein of human Transferrin (NP_001054) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI5G2 | |
| Flow Cytometry | |
| Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers | |
| 7018.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction