missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Transferrin Monoclonal Antibody (12A6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA120106
This item is not returnable.
View return policy
Description
MA1-20106 detects transferrin in human samples.
Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis.Serum transferrin may also have a further role in stimulating cell proliferation.
Specifications
| Transferrin | |
| Monoclonal | |
| 5.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P02787 | |
| Tf | |
| Placental transferrin receptor or transferrin. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 12A6 | |
| Unconjugated | |
| Tf | |
| AI266983; beta-1 metal-binding globulin; Cd176; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; HEL-S-71p; HP; hpx; hypotransferrinemia with hemochromatosis; ICA; Inhibitor of carbonic anhydrase; lactoferrin; liver regeneration-related protein LRRG03; liver transferrin; LOC100146633; PICA; porcine inhibitor of carbonic anhydrase; porcine inhibitor of carbonic anhydrase precursor; PRO1400; PRO1557; PRO2086; Serotransferrin; siderophilin; TF; Tfn; TFQTL1; transferin; Transferrin; transferrin precursor; Trf | |
| Mouse | |
| Protein A | |
| RUO | |
| 7018 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction