Learn More
Invitrogen™ Transferrin Receptor Monoclonal Antibody (R17217.1.4), PE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA517924
 View more versions of this product
Product Code. 15324201
  • € 712.00 / 300µg
Description
Transferrin Receptor Monoclonal Antibody for Flow
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transferrin Receptor
Monoclonal
0.1 mg/mL
PBS with 4-5mg/mL BSA and 0.02% sodium azide
Q62351
TFRC
Induced Friend Erythroleukemia 745.6 cell extracts.
300 μg
Primary
Mouse
Antibody
IgG2a
Flow Cytometry
R17217.1.4
PE
TFRC
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr
Rat
Protein G
RUO
22042
4°C, do not freeze