missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Transferrin Receptor Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5143016

Product Code. 17897632

  • € 474.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Transferrin Receptor
Polyclonal
Unconjugated
TFRC
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr
Goat
Ammonium sulfate precipitation
RUO
7037
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P02786
TFRC
Peptide with sequence TRFSLARQVDGDNSH-C (aa 21-35).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.