missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin/TF Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
€ 149.00 - € 401.60
Specifications
| Antigen | Transferrin |
|---|---|
| Clone | 5O7 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235465
|
Proteintech
81676-1-RR-20UL |
20 μL |
€ 149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233553
|
Proteintech
81676-1-RR-100UL |
100 μL |
€ 401.60
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Transferrin antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Transferrin. This antibody recognizes human, mouse, rat antigen. The Transferrin antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| Transferrin | |
| 1000 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 7018 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 5O7 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| DKFZp781D0156; PRO1557; PRO2086 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title