missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transgelin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10234-2-AP-20UL
This item is not returnable.
View return policy
Description
This antibody was generated against full length transgenlin 2 protein. It may cross react with other two transgelins based on the sequence similarity.
The protein encoded by this gene is a homolog of the protein transgelin, which is one of the earliest markers of differentiated smooth muscle. The function of this protein has not yet been determined.Specifications
| Transgelin 2 | |
| Polyclonal | |
| Unconjugated | |
| TAGLN2 | |
| SM22 beta, SM22beta, TAGLN2, Transgelin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21346, 8407 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P37802, Q9WVA4 | |
| TAGLN2 | |
| Transgelin 2 Fusion Protein Ag0337 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction