Learn More
Transglutaminase 2/TGM2 Antibody (013), DyLight 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89793UV
Product Code. 18560210
  • € 625.00 / 0.10mL
Description
Transglutaminase 2/TGM2 Monoclonal antibody specifically detects Transglutaminase 2/TGM2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transglutaminase 2/TGM2
Monoclonal
DyLight 350
C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2
This antibody was obtained from a rabbit immunized with purified, recombinant Human Transglutaminase 2/TGM2 (NP_004604.2; Met 1-Ala 687).
0.1 mL
Apoptosis, Immune System Diseases, Immunology, Signal Transduction
7052
Store at 4°C in the dark.
IgG
ELISA
013
50mM Sodium Borate
Rabbit
Protein A purified