missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 2/TGM2 Antibody (TGM2/6943R) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14193JF525
This item is not returnable.
View return policy
Description
Transglutaminase 2/TGM2 Monoclonal antibody specifically detects Transglutaminase 2/TGM2 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Transglutaminase 2/TGM2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGCGNAH, Tissue transglutaminase, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2 | |
| Recombinant full-length human Transglutaminase 2/TGM2 protein (Uniprot: P21980) | |
| 0.1 mL | |
| Apoptosis, Immune System Diseases, Immunology, Signal Transduction | |
| 7052 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| TGM2/6943R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction