missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 3/TGM3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97324CL1
This item is not returnable.
View return policy
Description
Transglutaminase 3/TGM3 Polyclonal antibody specifically detects Transglutaminase 3/TGM3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Transglutaminase 3/TGM3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| EC 2.3.2.13, MGC126250, protein-glutamine gamma-glutamyltransferase E, TG(E), TGase E, TGase-3, TGEMGC126249, transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase E, transglutaminase-3 | |
| Produced in rabbits immunized with E. coli-derived Human Transglutaminase 3/TGM3 fragment. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 7053 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction