missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transglutaminase 5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94524-0.1ml
This item is not returnable.
View return policy
Description
Transglutaminase 5 Polyclonal antibody specifically detects Transglutaminase 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Transglutaminase 5 | |
| Polyclonal | |
| Western Blot 1:2000 - 1:8000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| EC 2.3.2.13, protein-glutamine gamma-glutamyltransferase 5, TG(X), TGase X, TGase-5, TGASE5, TGASEX, TGM6, TGMX, TGXMGC141907, transglutaminase 5, transglutaminase V, Transglutaminase X, transglutaminase-5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 461-720 of human TGM5 (NP_963925.2). LQKLKARSFHGSQRGAELQPSRPTSLSQDSPRSLHTPSLRPSDVVQVSLKFKLLDPPNMGQDICFVLLALNMSSQFKDLKVNLSAQSLLHDGSPLSPFWQDTAFITLSPKEAKTYPCKISYSQYSQYLSTDKLIRISALGEEKSSPEKILVNKIITLSYPSITINVLGAAVVNQPLSIQVIFSNPLSEQVEDCVLTVEGSGLFKKQQKVFLGVLKPQHQASIILETVPFKSGQRQIQANMRSNKFKDIKGYRNVYVDFAL | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 9333 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction