missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transketolase Antibody (OTI5H3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74585AF647
This item is not returnable.
View return policy
Description
Transketolase Monoclonal specifically detects Transketolase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Transketolase | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TKT | |
| Full length recombinant protein of human Transketolase (NP_001055) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI5H3 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| EC 2.2.1.1, FLJ34765, TK, TKT1, transketolase | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 7086.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction