Learn More
Transketolase Mouse anti-Human, CoraLite 488, Clone: 3E2F3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66016-100UL
Description
The Transketolase antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Transketolase. This antibody recognizes human antigen. The Transketolase antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| Transketolase | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 7086 | |
| IgG2b |
| Immunofluorescence | |
| 3E2F3 | |
| Mouse | |
| Primary | |
| Human |