missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Transketolase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118312
This item is not returnable.
View return policy
Description
Transketolase Polyclonal Antibody for Western Blot, IP
Transketolase encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| Transketolase | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P29401 | |
| TKT | |
| E. coli-derived Human TKT fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| TKT | |
| epididymis luminal protein 107; epididymis secretory protein Li 48; HEL107; HEL-S-48; p68; TK; TKT; TKT1; Transketolase; transketolase (Wernicke-Korsakoff syndrome) | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7086 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction