missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Cellular tumor antigen p53 siRNA
Blocks transcription and translation of Cellular tumor antigen p53 from Human.
Brand: enQuireBio™ QX32-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_000537 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7157 | |
10 nmol | |
Human |
U-2 OS Huan Osteosarcoma Epithelial Cells from Bone. | |
TP53 | |
siRNA | |
TRP53, BCC7, P53, LFS1, TP53 | |
Lyophilized |