missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TLR9 / CD289 siRNA
Blocks transcription and translation of TLR9 / CD289 from Human.
Brand: enQuireBio™ QX28-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_059138 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
54106 | |
10 nmol | |
Human |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
TLR9 | |
siRNA | |
CD289, Toll-like receptor 9, TLR-9 | |
Lyophilized |