Learn More
Masterflex™ Translucent PTFE Microbore Tubing
Product Code. 12716278 Shop All Masterflex Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.042 mm
1.3208 mm
Diameter (Metric) Outer:
0.066 mm
1.9304 mm
Unit Size:
100ft
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Unit Size
12716278 1.3208 mm 1.9304 mm Each
15342437 0.042 mm 0.066 mm 100ft
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12716278 Supplier Masterflex™ Supplier No. 0641751
  • ONLINE EXCLUSIVE PRICE
    € 84.00
    € 73.37 / Pack of 1
    Save € 10.63
    12% Off
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
Cole-Parmer Translucent PTFE Microbore Tubing, Diameter Inner: 0.052 mm, Diameter Outer: 0.076 mm, For Use With: Used in extreme temperatures and in harsh chemical environments, Maximum Pressure: 4000 psi, Max. Operating Temperature: -240 deg.C to +260 deg.C, Min. Operating Temperature: -240 deg.C
Specifications
Diameter (English) Inner 0.053 in.
Diameter (Metric) Inner 1.3208 mm
Material PTFE
Diameter (English) Outer 0.076 in.
Diameter (Metric) Outer 1.9304 mm
For Use With (Application) Used in extreme temperatures and in harsh chemical environments
Min. Operating Temperature (Metric) -240°C
Packaging Quantity 1 Each
Color Translucent
Maximum Pressure 4000 psi
Max. Operating Temperature (English) 500°F
Max. Operating Temperature (Metric) -240°C to +260°C
Product Type Microbore Tubing