Learn More
B Braun™ Transofix™ Transfer Set
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
Description
- Dissolution of lypophilisates
- Mixing solutions and medicine
- Flooding of erythrocyte concentrate
- Slender, slightly siliconized spikes with round edges to prevent shearing of rubber particles
- Comfortable handle plate
Specifications
Specifications
| Color | Clear |
| Sterility | Sterile |