Learn More
Vitlab™ Transparent HDPE Large Funnel
Made from HDPE. Vitlab™ Transparent HDPE Large Funnel is suitable for filling large amounts of liquids.
Brand:  Vitlab™ 41794
Additional Details : Weight : 0.01000kg
Product Code. 10251591
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Transparent
  • Rapid flow due to 60° angle
  • Practical handle for hanging 
  • Optional accessories available
Specifications
Specifications
200mm
HDPE
HDPE filter
1300 mL
200 mm
Suitable for filling large amounts of liquids