Learn More
VITLAB™ Narrow Mouth LDPE Safety Wash Bottles
Work safely with potentially dangerous chemicals. VITLAB™ VITsafe™ Narrow Mouth LDPE Safety Wash Bottles feature a permanent safety imprint, color-coded VENT-CAP and precise spray control to eliminate drips and leaks and ensure safe handling.
Brand:  VITLAB™ 1331869
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 10511043
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
VITsafe™ Narrow Mouth LDPE Safety Wash Bottles – Wash bottles meet all safety requirements for working with potentially dangerous chemicals.
Specifications
Specifications
Safety Wash Bottle
58 mm
Ethanol
Transparent
Individually wrapped in PE bag
Orange
Narrow
135 mm
Vented Screw Cap
250 mL
LDPE, Polypropylene
12Pack
GL 25