Learn More
VITLAB™ Narrow Mouth LDPE Safety Wash Bottles
Work safely with potentially dangerous chemicals. VITLAB™ VITsafe™ Narrow Mouth LDPE Safety Wash Bottles feature a permanent safety imprint, color-coded VENT-CAP and precise spray control to eliminate drips and leaks and ensure safe handling.
Brand:  VITLAB™ 1332969
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10517292
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
VITsafe™ Narrow Mouth LDPE Safety Wash Bottles – Wash bottles meet all safety requirements for working with potentially dangerous chemicals.
Specifications
Specifications
Safety Wash Bottle
74 mm
Acetonitrile
Transparent
Individually wrapped in PE bag
Red
Narrow
180 mm
Vented Screw Cap
500 mL
LDPE, Polypropylene
6Pack
GL 25