Learn More
VITLAB™ Transparent LDPE Wide Mouth Wash Bottles
Click to view available options
:
Capacity: 250mL, GL 45 Thread Cap
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | LDPE |
| Mouth | Wide |
| Product Type | Wash Bottle |
| Closure Size | GL 45 |