Learn More
Deutsch Neumann™ Transparent Natural Rubber Latex Tubing
Natural Rubber Latex Tubing that is sterilizable at 120°C.
Brand: Deutsch Neumann™ 3051016
Additional Details : Weight : 1.15290kg
Description
- Hardness of 40 Shore A
- Light and flexible
- Offers excellent tensile strength, elongation, resilience and tear resistance
- Available in 10 m rolls
Specifications
ISO 9001:2008 | |
Natural Rubber | |
10 m | |
Non-sterile |
10 mm | |
16 mm | |
3 mm | |
Transparent |