Learn More
Deutsch Neumann™ Transparent Natural Rubber Latex Tubing
Natural Rubber Latex Tubing that is sterilizable at 120°C.
Brand:  Deutsch Neumann™ 3051217
Additional Details : Weight : 0.12000kg
 View more versions of this product
Product Code. 15292129
  • € 108.00 / 10m
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Description
Description
  • Hardness of 40 Shore A
  • Light and flexible
  • Offers excellent tensile strength, elongation, resilience and tear resistance
  • Available in 10 m rolls
Specifications
Specifications
ISO 9001:2008
Natural Rubber
10 m
Non-sterile
12 mm
17 mm
2.5 mm
Transparent