Learn More
VITLAB™ Transparent Polypropylene Sample Containers with Snap-On Cap
Click to view available options
Capacity (Metric):
18 mL
160 mL
Diameter (Metric):
22 mm
50 mm
Height (Metric):
57 mm
110 mm
Unit Size:
Each
Description
- Product Type: Sample Container
- Material: Polypropylene (Container), LDPE (Cap)
- Color: Transparent
- Shape: Round
- Includes: Snap-On Cap
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Graduated | No |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Capacity (English) | 0.6 oz. |
| Capacity (Metric) | 18 mL |
| Closure Material | LDPE |
| Closure Type | Snap Cap |
| Color | Transparent |
| Show More |