Learn More
VITLAB™ Wash Bottles, Transparent Polypropylene
Product Code. 10651965
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Thread Size (Metric):
GL 25
GL 32
GL 45
GL 63
Unit Size:
Each
Pack of 12
Product Code. 10651965
Brand: VITLAB™ 93793
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Wash equipment and materials with a precise spray jet and optimized backflow. VITLAB™ Wash Bottles, Transparent Polypropylene feature a screw cap with an incorporated spray tube on a narrow- or wide-mouth bottle. Spray bottles are individually wrapped, autoclavable and recyclable.
Wash bottles are appropriate for cleaning, coating and dousing in any laboratory environment. Polypropylene bottles are ideal for a variety of applications, including use with a range of chemicals, solvents and temperatures. Spray bottles are individually wrapped in clearly labeled polyethylene bags.
  • Designed for precise spray control
    • Finely drawn tip creates a precise spray jet for fine control
    • Optimised medium backflow through the continuous spray tube
  • Choice of narrow- or wide-mouth bottles
    • Narrow-mouth, tapered bottles for easier handling
    • Wide-mouth, square-shouldered bottles for easy and safe filling
    • All bottles feature screw cap lids with GL threads for a strong, secure hold
  • Constructed of durable, transparent polypropylene
    • Chemically resistant for use with liquid or powdered reagents
    • Temperature resistant and autoclavable
    • Transparent material for easy identification and monitoring
Specifications
Capacity (Metric) 250 mL
Color Transparent
Material Polypropylene
Quantity 12Pack
Thread Size (Metric) GL 45
Mouth Wide
Height (Metric) 146 mm
Product Type Wash Bottle
Diameter (Metric) 58 mm
Closure Type Screw Cap