missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transportin 1 Antibody (D45), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1397H
This item is not returnable.
View return policy
Description
Transportin 1 Monoclonal specifically detects Transportin 1 in Human, Mouse, Bovine, Canine, Monkey, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Transportin 1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TNPO1 | |
| Human recombinant His-tagged Transportin 1. [UniProt# Q92973] | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 3842 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| D45 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| importin beta 2, Importin beta-2, IPO2, karyopherin (importin) beta 2, MIP1Karyopherin beta-2, MIPM9 region interaction protein, transportin 1, transportin-1, TRNKPNB2importin 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Canine, Monkey, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction