missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transthyretin/Prealbumin Antibody (TTR/4292), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08765B
This item is not returnable.
View return policy
Description
Transthyretin/Prealbumin Monoclonal antibody specifically detects Transthyretin/Prealbumin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Transthyretin/Prealbumin | |
| Monoclonal | |
| Biotin | |
| ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin | |
| Recombinant human full-length protein (Uniprot: P02766) | |
| 100 μg | |
| Cancer | |
| 7276 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| TTR/4292 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction