missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Transthyretin/Prealbumin Native Protein

Product Code. 18629047 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18629047

Brand: Novus Biologicals™ NBP262523

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Formulation 0.02 M Ammonium bicarbonate (NH4HCO3)
Gene ID (Entrez) 7276
Molecular Weight (g/mol) MolecularWeight-theroretical: 60 kDa
Quantity 1 mg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias ATTR, HsT2651, PALBTBPA, Prealbumin, prealbumin, amyloidosis type I, thyroxine-binding prealbumin, transthyretin
Gene Symbol TTR
Reconstitution Reconstitute with 1 mL sterile PBS. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. Gently mix vial after reconstitution.)
Research Category Cancer
Purity or Quality Grade >95%, by SDS-PAGE
Protein Transthyretin/Prealbumin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.