missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAP1 Antibody (3H4-2H6), Novus Biologicals™
Mouse Monoclonal Antibody
€ 166.00
Specifications
| Antigen | TRAP1 |
|---|---|
| Clone | 3H4-2H6 |
| Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
Description
TRAP1 Monoclonal specifically detects TRAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| TRAP1 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 10131 | |
| Purified recombinant TRAP1 | |
| Primary | |
| Detects 75kDa. |
| 3H4-2H6 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein | |
| TRAP1 | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title