missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47598
This item is not returnable.
View return policy
Description
TRAP1 Polyclonal specifically detects TRAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TRAP1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| HSP 75, HSP75TNFR-associated protein 1, HSP90Lheat shock protein 75 kDa, mitochondrial, TNF receptor-associated protein 1, TRAP-1, tumor necrosis factor type 1 receptor associated protein, Tumor necrosis factor type 1 receptor-associated protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TRAP1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RTTAQLGPRRNPAWSLQAGRLFSTQTAEDKEEPLHSIISSTESVQGSTSKHEFQAETKKLLDIVARSLYSEKEVFIREL | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 10131 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction