missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12484-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is thought to be part of a large multi-subunit complex involved in the targeting and fusion of endoplasmic reticulum-to-Golgi transport vesicles with their acceptor compartment. In addition, the encoded protein can bind c-myc promoter-binding protein 1 and block its transcriptional repression capability. Mutations in this gene are a cause of spondyloepiphyseal dysplasia tarda (SEDT). A processed pseudogene of this gene is located on chromosome 19, and other pseudogenes are found on chromosomes 8 and Y. Alternatively spliced transcript variants have been found for this gene.Specifications
| TRAPPC2 | |
| Polyclonal | |
| Unconjugated | |
| TRAPPC2 | |
| hYP38334, MBP 1 interacting protein 2A, MIP 2A, SEDL, Sedlin, SEDLP, SEDT, TRAPPC2, TRAPPC2P1, TRS20, ZNF547L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 501550, 6399, 66226 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| D3ZVF4, P0DI81, Q9CQP2 | |
| TRAPPC2 | |
| TRAPPC2 Fusion Protein Ag3201 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction