missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC2L Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98640H
This item is not returnable.
View return policy
Description
TRAPPC2L Polyclonal antibody specifically detects TRAPPC2L in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TRAPPC2L | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 51693 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| HRP | |
| MGC111156, trafficking protein particle complex 2-like, trafficking protein particle complex subunit 2-like protein | |
| Produced in rabbits immunized with E. coli-derived Human TRAPPC2L fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction