missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRAPPC2L Polyclonal Antibody
GREENER_CHOICE

Product Code. 17111394 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17111394 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17111394 Supplier Invitrogen™ Supplier No. PA5113531

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRAPPC2L Polyclonal Antibody for Western Blot, IHC (P), ELISA

May play a role in vesicular transport from endoplasmic reticulum to Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC2L
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TRAPPC2L
Gene Accession No. Q9JME7, Q9UL33
Gene Alias 1810017G16Rik; AB030200; hematopoietic stem/progenitor cells 176; HSPC126; HSPC176; RGD1304906; Tca17; trafficking protein particle complex 2-like; trafficking protein particle complex subunit 2-like protein; TRAPPC2L
Gene Symbols TRAPPC2L
Host Species Rabbit
Immunogen Recombinant Human Trafficking protein particle complex subunit 2-like protein (1-139AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51693, 59005
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.