missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAPPC4 Monoclonal Antibody (OTI4B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505629
This item is not returnable.
View return policy
Description
TRAPPC4 is part of the multisubunit TRAPP (transport protein particle) complex and interacts with SDC2. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPP proteins are involved in tethering during vesicle transport.Specifications
| TRAPPC4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y296 | |
| TRAPPC4 | |
| Full length human recombit protein of human TRAPPC4 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4B5 | |
| Unconjugated | |
| TRAPPC4 | |
| CGI-104, HSPC172, PTD009, SBDN, SYNBINDIN, TRS23 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51399 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction