missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAPPC6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560212
This item is not returnable.
View return policy
Description
Immunogen sequence: VGQALGERLP RETLAFREEL DVLKFLCKDL WVAVFQKQMD SLR Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 86%.
TRAPPC6A is a subunit of the TRAPP I and II complexes.
Specifications
| TRAPPC6A | |
| Polyclonal | |
| Unconjugated | |
| TRAPPC6A | |
| HSPC289; trafficking protein particle complex 6A; trafficking protein particle complex subunit 6A; TRAPP complex subunit 6A; TRAPPC6A; TRAPPC6Adelta29-42; TRS33 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79090 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75865 | |
| TRAPPC6A | |
| Recombinant protein corresponding to Human TRAPPC6A. Recombinant protein control fragment (Product #RP-98855). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction